Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Puradisc 30 syringe filters
Products
Product Category
syringe filters (20)
Color
Manufacturer
Diameter (mm)
Surface Area cm²
Volume (ml)
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
WHA10462200
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, sterile, 50/pk
Expand
View Pricing
WHA10462100
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.45 μm, sterile, 50/pk
Expand
View Pricing
WHA10462300
Whatman
®
Puradisc 30 syringe filters,
Puradisc FP 30, PTFE, 0.2 μm, sterile, 50/pk
Expand
View Pricing
WHA10462000
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose nitrate, 5.0 μm, sterile, 50/pk
Expand
View Pricing
WHA10462260
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate 1.2 μm, sterile, 50/pk
Expand
View Pricing
WHA10462205
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, luer-lock outlet, sterile, 50/pk
Expand
View Pricing
WHA10462261
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 1.2 μm, 50/pk
Expand
View Pricing
WHA10462701
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, 50/pk
Expand
View Pricing
WHA10462600
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.45 μm, 500/pk
Expand
View Pricing
WHA10462520
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose nitrate, 5.0 μm, 50/pk
Expand
View Pricing
WHA10462500
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose nitrate, 5.0 μm, 500/pk
Expand
View Pricing
WHA10462510
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose nitrate, 5.0 μm, 100/pk
Expand
View Pricing
WHA10462241
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.8 μm, 50/pk
Expand
View Pricing
WHA10462240
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.8 μm, sterile, 50/pk
Expand
View Pricing
WHA10462610
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.45 μm, 100/pk
Expand
View Pricing
WHA10462206
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, luer-lock outlet, 500/pk
Expand
View Pricing
WHA10462700
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, 500/pk
Expand
View Pricing
WHA10462263
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 1.2 μm, 500/pk
Expand
View Pricing
WHA10462243
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.8 μm, 500/pk
Expand
View Pricing
WHA10462710
Whatman
®
Puradisc 30 syringe filters,
Puradisc 30 syringe filter, cellulose acetate, 0.2 μm, 100/pk
Expand
View Pricing