Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V
Products
Product Category
filter discs (6)
Manufacturer
Diameter (mm)
Package Size
Pore size (µm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
WHA1204125
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 125 mm, pack of 100
Expand
View Pricing
WHA1204150
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 15 cm, pack of 100
Expand
View Pricing
WHA12040185
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 18.5 mm, pack of 100
Expand
View Pricing
WHA1204320
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 32 cm, pack of 100
Expand
View Pricing
WHA1204270
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 27 mm, pack of 14
Expand
View Pricing
WHA1204240
Whatman
®
Prepleated Qualitative Filter Paper, Grade 4V,
circles, diam. 24 cm, pack of 100
Expand
View Pricing