
Whatman® mixed cellulose ester WME filter discs 0.45 μm pore size

Synonym(s):
Whatman® nitrocellulose membrane filters
Products
Product Category
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Sterility
Showing 1-7 of 7
Compare
Product Number
Description
Pricing