Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
glass microfiber filters, Grade GF/B
Products
Product Category
filter discs (3)
Manufacturer
Length (cm)
Length (mm)
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
WHA1821915
Whatman
®
glass microfiber filters, Grade GF/B,
Grade GF/B sheets, 460 x 570 mm, 25/pk
Expand
View Pricing
WHA1821271
Whatman
®
glass microfiber filters, Grade GF/B,
sheets, L × W 2.25 in. × 12.25 in.
Expand
View Pricing
WHA1821914
Whatman
®
glass microfiber filters, Grade GF/B,
sheets, L × W 460 mm × 570 mm
Expand
View Pricing