Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vanadium nitride
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF54609015
Vanadium nitride,
powder, max. particle size 150 micron, weight 10 g, purity 99%
Expand
View Pricing
GF26127255
Vanadium nitride,
powder, max. particle size 150 micron, weight 20 g, purity 99%
Expand
View Pricing
GF38310128
Vanadium nitride,
powder, max. particle size 150 micron, weight 100 g, purity 99%
Expand
View Pricing
GF65906488
Vanadium nitride,
powder, 150 max. part. size (micron), weight 50 g, purity 99%
Expand
View Pricing
GF88766230
Vanadium nitride,
powder, 150 max. part. size (micron), weight 200 g, purity 99%
Expand
View Pricing