
Uridine 5′-diphosphoglucuronic acid trisodium salt

Synonym(s):
UDP-GlcA, UDPGA, Uridine-diphosphate-glucuronic acid trisodium salt, Uridine[5′]diphospho[1]-α-D-glucopyranosuronic acid trisodium salt
Empirical Formula (Hill Notation):
C15H19N2Na3O18P2
CAS Number:
Molecular Weight:
646.23
Products
Brand
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing