
TWEEN® 20

Synonym(s):
Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, TWEEN® 20
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Feature
Physical Form
Product Line
Chemical Composition
Quality Segment
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing