
Tryptic Soy Broth - Dehydrated Culture Media

Synonym(s):
CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth
Products
Product Category
Brand
Biological Source
Color
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing