Tryptamine hydrochloride
Synonym(s):
2-(3-Indolyl)ethylamine hydrochloride, 3-(2-Aminoethyl)indole hydrochloride
Empirical Formula (Hill Notation):
C10H12N2 · HCl
CAS Number:
Molecular Weight:
196.68
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2