Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TrueNorth
®
Cardboard Cryovial Box and Lid
Products
Product Category
boxes (6)
Color
Manufacturer
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
HS2860A
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 2 in., white, pk of 12
Expand
View Pricing
HS2860CR
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 2 in., red, pk of 12
Expand
View Pricing
HS2860C
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 2 in., assorted colors, pk of 12
Expand
View Pricing
HS2860B
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 3 in., white, pk of 12
Expand
View Pricing
HS2860CB
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, for tubes, 1.5 ‑ 2 mL, L × W × H 5.2 × 5.2 × 2 in., blue, pk of 12
Expand
View Pricing
HS2860CY
TrueNorth
®
Cardboard Cryovial Box and Lid,
without dividers, Holds 1.5-2 mL tubes, L × W × H 5.2 × 5.2 × 2 in., yellow, pk of 12
Expand
View Pricing