
Trospium chloride

Synonym(s):
(1α,3β,5α)-3-[(hydroxydiphenylacetyl)oxy]spiro[8-azoniabicyclo[3.2.1]octane-8,1’-pyrrolidinium] chloride, Azoniaspiro(3α-benziloyloxynortropane-8,1′-pyrrolidine) chloride
Empirical Formula (Hill Notation):
C25H30ClNO3
CAS Number:
Molecular Weight:
427.96
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-4 of 4