Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trizma
®
hydrochloride solution
Synonym(s):
Tris hydrochloride solution
EC Number:
234-318-8
Products
Product Category
buffers (12)
Brand
Sigma-Aldrich (12)
Feature
pH Value
Physical Form
Product Line
Quality Segment
Special Grade
BioPerformance Certified (10)
Biotechnology Performance Certified (10)
Sterilization Method
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
T2194
Trizma
®
hydrochloride solution,
pH 7.4, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T2694
Trizma
®
hydrochloride solution,
pH 8.0, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T3038
Trizma
®
hydrochloride solution,
1 M, BioReagent, for molecular biology
Expand
View Pricing
T2663
Trizma
®
hydrochloride solution,
1 M, BioReagent, for molecular biology
Expand
View Pricing
T2819
Trizma
®
hydrochloride solution,
pH 9.0, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T2319
Trizma
®
hydrochloride solution,
pH 7.5, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T1819
Trizma
®
hydrochloride solution,
pH 7.0, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T3069
Trizma
®
hydrochloride solution,
pH 8.0, BioPerformance Certified, 2 M, suitable for cell culture
Expand
View Pricing
T2444
Trizma
®
hydrochloride solution,
pH 7.6, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T2569
Trizma
®
hydrochloride solution,
pH 7.8, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T2069
Trizma
®
hydrochloride solution,
pH 7.2, BioPerformance Certified, 1 M, suitable for cell culture
Expand
View Pricing
T2944
Trizma
®
hydrochloride solution,
BioPerformance Certified, suitable for cell culture, pH 7.5
Expand
View Pricing