
Triton™ X-45

Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol 4-tert-octylphenyl ether
Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= ~5
CAS Number:
Products
Product Category
Brand
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing