Skip to Content
Merck
Triphosgene reagent grade, 98%

Triphosgene

Triphosgene reagent grade, 98%
Synonym(s):
Bis(trichloromethyl) carbonate
Linear Formula:
Cl3COCOOCCl3
CAS Number:
Molecular Weight:
296.75
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing