
Triphenylphosphine-3,3′,3′′-trisulfonic acid trisodium salt

Synonym(s):
3,3′,3″-Phosphinidynetris(benzenesulfonic acid) trisodium salt, Tris(3-sulfophenyl)phosphine trisodium salt
Linear Formula:
P(C6H4SO3Na)3
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing