
Trioxsalen

Synonym(s):
2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one, 4,5′,8-Trimethylpsoralen, TMP, Trisoralen
Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing