Skip to Content
Merck
Trimethyl(trifluoromethyl)silane solution 2 M in THF

Trimethyl(trifluoromethyl)silane solution

Trimethyl(trifluoromethyl)silane solution 2 M in THF
Synonym(s):
(Trifluoromethyl)trimethylsilane, Ruppert′s reagent, Ruppert-Prakash reagent, TFMTMS, Trifluoromethyltrimethylsilane
Linear Formula:
(CH3)3SiCF3
CAS Number:
Molecular Weight:
142.19

Products

Brand
Formula Weight
Physical Form
Functional Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing