Skip to Content
Merck
Triethyl phosphate ReagentPlus®, ≥99.8%

Triethyl phosphate

Triethyl phosphate ReagentPlus®, ≥99.8%
Synonym(s):
Ethyl phosphate, Triethyl phosphate
Linear Formula:
(C2H5O)3PO
CAS Number:
Molecular Weight:
182.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing