Skip to Content
Merck
Triethylenetetramine ≥97.0% (T)

Triethylenetetramine

Triethylenetetramine ≥97.0% (T)
Synonym(s):
DEH 24, EPH 925, Triethylenetetramine
Linear Formula:
(H2NCH2CH2NHCH2)2
CAS Number:
Molecular Weight:
146.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3