Skip to Content
Merck
Tridecanal technical grade, 90%

Tridecanal

Tridecanal technical grade, 90%
Synonym(s):
Tridecyl aldehyde
Linear Formula:
CH3(CH2)11CHO
CAS Number:
Molecular Weight:
198.34
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing