
Trichostatin A

Synonym(s):
TSA, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide, Trichostatin A, Streptomyces sp.
Empirical Formula (Hill Notation):
C17H22N2O3
CAS Number:
Molecular Weight:
302.37
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-2 of 2