Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trichlorofluoromethane solution
CAS Number:
75-69-4
Products
Product Category
stable isotopes (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Physical Form
Format
Chemical Composition
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
710717
Trichlorofluoromethane solution,
NMR reference standard, 10% in acetone-d
6
(99.9 atom % D), TMS 6 %, 1,2-dichloro-1,1-difluoroethane 1 %
Expand
View Pricing
612715
Trichlorofluoromethane solution,
NMR reference standard, 50% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
Expand
View Pricing