
Tributylphosphine oxide

Synonym(s):
Butyphos, Tributylphosphine oxide
Linear Formula:
[CH3(CH2)3]3P(O)
CAS Number:
Molecular Weight:
218.32
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reaction Type
Reagent Type
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing