
trans-p-Coumaric acid

Synonym(s):
p-Coumaric acid, trans-4-Hydroxycinnamic acid
Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing