Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(−)-
trans
-Caryophyllene
Synonym(s):
β-Caryophyllene,
trans
-(1
R
,9
S
)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene
Empirical Formula (Hill Notation):
C
15
H
24
CAS Number:
87-44-5
Molecular Weight:
204.35
EC Number:
201-746-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (2)
Puriss (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
22075
(−)-
trans
-Caryophyllene,
≥98.0% (sum of enantiomers, GC)
Expand
View Pricing
PHL80717
(−)-
trans
-Caryophyllene,
phyproof
®
Reference Substance
Expand
View Pricing
75541
(−)-
trans
-Caryophyllene,
analytical standard
Expand
View Pricing