Skip to Content
Merck
trans-2-Octenal analytical standard

trans-2-Octenal

trans-2-Octenal analytical standard
Linear Formula:
CH3(CH2)4CH=CHCHO
CAS Number:
Molecular Weight:
126.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing