
Torsemide Related Compound A

Synonym(s):
4-[(3-Methylphenyl)amino]-3-pyridinesulfonamide
Empirical Formula (Hill Notation):
C12H13N3O2S
CAS Number:
Molecular Weight:
263.32
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing