Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Toluidine Blue O
Synonym(s):
Basic Blue 17, Blutene chloride, Methylene Blue T50 or T extra, Tolonium chloride
Empirical Formula (Hill Notation):
C
15
H
16
ClN
3
S
CAS Number:
92-31-9
Molecular Weight:
305.83
EC Number:
202-146-2
Products
Product Category
stains (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T3260
Toluidine Blue O,
Powder
Expand
View Pricing
198161
Toluidine Blue O,
certified by the Biological Stain Commission
Expand
View Pricing