
Tolbutamide

Synonym(s):
1-Butyl-3-(4-methylphenylsulfonyl)urea, N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide
Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Products
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3