
Titanium(IV) chloride tetrahydrofuran complex

Synonym(s):
Tetrachlorobis(tetrahydrofuran)titanium
Linear Formula:
TiCl4 · 2THF
CAS Number:
Molecular Weight:
333.89
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing