Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tin(IV) oxide
Synonym(s):
Stannic oxide, Tin(IV) oxide
Linear Formula:
SnO
2
CAS Number:
18282-10-5
Molecular Weight:
150.71
Products
Brand
Sigma-Aldrich (3)
Supelco (1)
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
549657
Tin(IV) oxide,
nanopowder, ≤100 nm avg. part. size
Expand
View Pricing
204714
Tin(IV) oxide,
≥99.99% trace metals basis
Expand
View Pricing
244651
Tin(IV) oxide,
−325 mesh, 99.9% trace metals basis
Expand
View Pricing
1.07818
Tin(IV) oxide,
EMPLURA
®
Expand
View Pricing