
Thyrotropic hormone from bovine pituitary

Synonym(s):
TSH, Thyroid stimulating hormone, Thyrotropin from bovine pituitary glands
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Tissue
Package Size
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing