Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thymolphthalein
Synonym(s):
Thymolphthalein, 5′,5′′-Diisopropyl-2′,2′′-dimethylphenolphthalein
Empirical Formula (Hill Notation):
C
28
H
30
O
4
CAS Number:
125-20-2
Molecular Weight:
430.54
EC Number:
204-729-7
Products
Product Category
dyes (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.08175
Thymolphthalein,
pH indicator, ACS, Reag. Ph Eur
Expand
View Pricing
89360
Thymolphthalein,
ACS reagent, indicator (pH 8.8-10.5)
Expand
View Pricing
114553
Thymolphthalein,
ACS reagent, Dye content 95 %
Expand
View Pricing