Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thymol blue
Synonym(s):
Thymol blue, Thymolsulfonphthalein
Empirical Formula (Hill Notation):
C
27
H
30
O
5
S
CAS Number:
76-61-9
Molecular Weight:
466.59
EC Number:
200-973-3
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Agency
Quality Segment
Application
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.08176
Thymol blue,
pH indicator, ACS, Reag. Ph Eur
Expand
View Pricing
114545
Thymol blue,
ACS reagent
Expand
View Pricing