Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thymine(Zidovudine Related Compound C)
Synonym(s):
Thymine, 2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil
Empirical Formula (Hill Notation):
C
5
H
6
N
2
O
2
CAS Number:
65-71-4
Molecular Weight:
126.11
EC Number:
200-616-1
Products
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHR1345
Thymine(Zidovudine Related Compound C),
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing