Skip to Content
Merck
Thiourea Pharmaceutical Secondary Standard; Certified Reference Material

Thiourea

Thiourea Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Sulfourea, Thiocarbamide, Thiourea
Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Reagent Type
API Family
Reference Material Type
Special Grade
Technique