Skip to Content
Merck
Thiazole Orange Dye content ~90 %

Thiazole Orange

Thiazole Orange Dye content ~90 %
Synonym(s):
1-Methyl-4-[(3-methyl-2(3H)-benzothiazolylidene)methyl]quinolinium p-tosylate
Empirical Formula (Hill Notation):
C26H24N2O3S2
CAS Number:
Molecular Weight:
476.61

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing