Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thermolysin from
Geobacillus stearothermophilus
Synonym(s):
Protease from
Geobacillus stearothermophilus
, Thermophilic-bacterial protease
CAS Number:
9073-78-3
EC Number:
232-973-4
Products
Product Category
proteases (1)
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P1512
Thermolysin from
Geobacillus stearothermophilus
,
Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)
Expand
View Pricing
T7902
Thermolysin from
Geobacillus stearothermophilus
,
powder, BioReagent, 30-350 units/mg protein (E1%/280), suitable for cell culture
Expand
View Pricing