Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ThalesNano MidiCart
™
catalyst cartridge system
EC Number:
231-115-6
Products
Manufacturer
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
THSD2145
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Ru/C
Expand
View Pricing
THSD1146
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, Raney cobalt
Expand
View Pricing
THSD1142
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, Raney nickel
Expand
View Pricing
THSD1141
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 10% Pd/C
Expand
View Pricing
THSD1040
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, inert quartz
Expand
View Pricing
THSD2142
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Ru/Al
2
O
3
Expand
View Pricing
THSD1140
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, inert titanium
Expand
View Pricing
THSD2148
ThalesNano MidiCart
™
catalyst cartridge system,
5% Rh/AL
2
O
3
, L × diam. 116 cm × 9.2 mm
Expand
View Pricing
THSD1444
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 1%Pt/SiO
2
Expand
View Pricing
THSD1143
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 10% Pt/C
Expand
View Pricing
THSD1144
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Rh/C
Expand
View Pricing
THSD1148
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Pd/Al
2
O
3
Expand
View Pricing
THSD4144
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5%Pd/SiO
2
Expand
View Pricing
THSD1447
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, nickel sponge
Expand
View Pricing
THSD2141
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Pd/C
Expand
View Pricing
THSD2143
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Pt/Al
2
O
3
Expand
View Pricing
THSD3144
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, nickel sponge, 1% Mo
Expand
View Pricing
THSD1445
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 1%Pt/SiO
2
/polyethyleneimine
Expand
View Pricing
THSD6145
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 1% Pt/C
Expand
View Pricing
THSD8142
ThalesNano MidiCart
™
catalyst cartridge system,
L × diam. 116 mm × 9.2 mm, 5% Pt/C
Expand
View Pricing
Page 1 of 2
Page 1 of 2