Skip to Content
Merck
Tetraethylene glycol di(p-toluenesulfonate) 97%

Tetraethylene glycol di(p-toluenesulfonate)

Tetraethylene glycol di(p-toluenesulfonate) 97%
Synonym(s):
Tetraethylene glycol ditosylate
Linear Formula:
(CH3C6H4SO3CH2CH2OCH2CH2)2O
CAS Number:
Molecular Weight:
502.60

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing