
Tetracaine hydrochloride

Synonym(s):
4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester, Amethocaine hydrochloride
Empirical Formula (Hill Notation):
C15H24N2O2 · HCl
CAS Number:
Molecular Weight:
300.82
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing