
Tetracaine

Synonym(s):
4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester
Empirical Formula (Hill Notation):
C15H24N2O2
CAS Number:
Molecular Weight:
264.36
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3