Skip to Content
Merck
Tetrabutylammonium hydroxide solution 53.5-56.5% in H2O

Tetrabutylammonium hydroxide solution

Tetrabutylammonium hydroxide solution 53.5-56.5% in H2O
Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Functional Group
Special Grade
Technique