Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TDS
3
™
Storage Container
Products
Brand
Supelco (4)
Sustainability
Feature
Length (cm)
Compatibility
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
25097-U
TDS
3
™
Storage Container,
for use with PerkinElmer ATD-50, ATD-400, Turbomatrix, for use with Markes International Unity
Expand
View Pricing
25095-U
TDS
3
™
Storage Container,
for use with Tekmar
®
AEROtrap 6000, for use with Gerstel
®
TDS 2/TDS/A
Expand
View Pricing
28307-U
TDS
3
™
Storage Container,
for use with Gerstel 60mm TDU Tubes
Expand
View Pricing
25096-U
TDS
3
™
Storage Container,
for use with Dynatherm
™
850/890, ACEM 900/901-FF (Standard sampling tube)
Expand
View Pricing