
Tamoxifen citrate

Synonym(s):
Tamoxifen citrate salt, (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene
Empirical Formula (Hill Notation):
C26H29NO · C6H8O7
CAS Number:
Molecular Weight:
563.64
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing