
Tadalafil

Synonym(s):
Tadalafil
Empirical Formula (Hill Notation):
C22H19N3O4
CAS Number:
Molecular Weight:
389.40
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Technique