
Sulfanilamide

Synonym(s):
Sulfanilamide, p-Aminobenzenesulfonamide
Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Products
Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing