
Sudan I-d5

Synonym(s):
1-Phenylazo-2-naphthol-d5
Linear Formula:
C6D5N=NC10H6OH
CAS Number:
Molecular Weight:
253.31
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing