
Sucrose-13C12

Empirical Formula (Hill Notation):
13C12H22O11
CAS Number:
Molecular Weight:
354.21
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing