
Substance P acetate salt hydrate

Empirical Formula (Hill Notation):
C63H98N18O13S · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
1347.63 (anhydrous free base basis)
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing